accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 591-12-8

IUPAC Name: 5-methyl-2,3-dihydrofuran-2-one
Synonyms: 5-methyl-2(3H)-furanone delta(2)-angelica lactone α-angelica lactone 5-methylfuranone delta(sup 2)-angelica lactone α-angelicalactone β,gamma-angelica lactone α-angelicalacton
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: QOTQFLOTGBBMEX-UHFFFAOYSA-N
SMILES: CC1=CCC(=O)O1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)