missing translation for 'onlineSavingsMsg'
Learn More

alpha-Angelicalactone, 98%

Catalog No. AAL0760206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0760206 5 g
AAL0760214 25 g
AAL0760222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0760206 Supplier Thermo Scientific Chemicals Supplier No. L0760206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 591-12-8 | C5H6O2 | 98.10 g/mol

alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.

Solubility
Soluble in water (5g/100mL) and alcohol.

Notes
Moisture-sensitive. Store under inert gas. Protect from moisture. Incompatible materials are Oxidizing agents.

Chemical Identifiers

CAS 591-12-8
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00005375
InChI Key QOTQFLOTGBBMEX-UHFFFAOYSA-N
Synonym alpha-angelica lactone, alpha-angelicalactone, 5-methyl-2 3h-furanone, 5-methylfuran-2 3h-one, 2 3h-furanone, 5-methyl, 4-hydroxypent-3-enoic acid lactone, delta 2-angelica lactone, 4-hydroxy-3-pentenoic acid gamma-lactone, gamma-methyl-beta,gamma-crotonolactone, .alpha.-angelica lactone
PubChem CID 11559
ChEBI CHEBI:36433
IUPAC Name 5-methyl-3H-furan-2-one
SMILES CC1=CCC(=O)O1

Specifications

Melting Point 17°C to 18°C
Density 1.093
Boiling Point 53°C to 54°C (10 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.448
Quantity 5 g
Beilstein 108394
Merck Index 14,647
Solubility Information Soluble in water (5g/100mL) and alcohol.
Formula Weight 98.1
Percent Purity 98%
Chemical Name or Material alpha-Angelicalactone
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 209-701-8
RTECSNumber LU5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only