missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dimethylphenylacetic acid

CAS: 826-55-1 | C10H12O2 | 164.20 g/mol

$220.66 - $741.42

Chemical Identifiers

CAS 826-55-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00014332
InChI Key YYEROYLAYAVZNW-UHFFFAOYSA-N
Synonym 2-methyl-2-phenylpropionic acid, 2-phenylisobutyric acid, alpha,alpha-dimethylphenylacetic acid, unii-dr8huh2gyk, dimethylphenylacetic acid, dr8huh2gyk, 2-methyl-2-phenyl-propionic acid, hydratropic acid, .alpha.-methyl, benzeneacetic acid,.alpha.-dimethyl, propionic acid, 2-methyl-2-phenyl
PubChem CID 13222
IUPAC Name 2-methyl-2-phenylpropanoic acid
SMILES CC(C)(C(O)=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AA3288404
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
03288404
2 g
Each for $220.66
Only null left
 
AA3288409
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
03288409
10 g
Each for $741.42
Only null left
 
Description

Description

α,α-Dimethylphenylacetic acid is used in the synthesis of active pharmaceutical ingredient, fexofenadine hydrochloride, which is used as antihistamine. Further, it acts as pharmaceutical intermediates and as chemical intermediates in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

826-55-1
164.20
YYEROYLAYAVZNW-UHFFFAOYSA-N
13222
CC(C)(C(O)=O)C1=CC=CC=C1
C10H12O2
MFCD00014332
2-methyl-2-phenylpropionic acid, 2-phenylisobutyric acid, alpha,alpha-dimethylphenylacetic acid, unii-dr8huh2gyk, dimethylphenylacetic acid, dr8huh2gyk, 2-methyl-2-phenyl-propionic acid, hydratropic acid, .alpha.-methyl, benzeneacetic acid,.alpha.-dimethyl, propionic acid, 2-methyl-2-phenyl
2-methyl-2-phenylpropanoic acid

Specifications

826-55-1
146.5°C
MFCD00014332
2-methyl-2-phenylpropionic acid, 2-phenylisobutyric acid, alpha,alpha-dimethylphenylacetic acid, unii-dr8huh2gyk, dimethylphenylacetic acid, dr8huh2gyk, 2-methyl-2-phenyl-propionic acid, hydratropic acid, .alpha.-methyl, benzeneacetic acid,.alpha.-dimethyl, propionic acid, 2-methyl-2-phenyl
YYEROYLAYAVZNW-UHFFFAOYSA-N
2-methyl-2-phenylpropanoic acid
13222
Crystalline
80°C to 82°C
C10H12O2
2 g
Insoluble in water.
CC(C)(C(O)=O)C1=CC=CC=C1
164.20
164.21
alpha,alpha-Dimethylphenylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 212-556-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only