missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dimethylphenylacetic acid

Catalog No. AA3288409
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3288409 10 g
AA3288404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3288409 Supplier Thermo Scientific Chemicals Supplier No. 03288409

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 826-55-1 | C10H12O2 | 164.20 g/mol

α,α-Dimethylphenylacetic acid is used in the synthesis of active pharmaceutical ingredient, fexofenadine hydrochloride, which is used as antihistamine. Further, it acts as pharmaceutical intermediates and as chemical intermediates in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 826-55-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00014332
InChI Key YYEROYLAYAVZNW-UHFFFAOYSA-N
Synonym 2-methyl-2-phenylpropionic acid, 2-phenylisobutyric acid, alpha,alpha-dimethylphenylacetic acid, unii-dr8huh2gyk, dimethylphenylacetic acid, dr8huh2gyk, 2-methyl-2-phenyl-propionic acid, hydratropic acid, .alpha.-methyl, benzeneacetic acid,.alpha.-dimethyl, propionic acid, 2-methyl-2-phenyl
PubChem CID 13222
IUPAC Name 2-methyl-2-phenylpropanoic acid
SMILES CC(C)(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 80°C to 82°C
Boiling Point 146.5°C
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 164.21
Physical Form Crystalline
Chemical Name or Material alpha,alpha-Dimethylphenylacetic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 212-556-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only