missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dichlorodiphenylmethane 98.0+%, TCI America™

Catalog No. D036125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D036125G 25 g
D0361250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D036125G Supplier TCI America Supplier No. D036125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2051-90-3
Molecular Formula C13H10Cl2
Molecular Weight (g/mol) 237.123
MDL Number MFCD00000811
InChI Key OPTDDWCXQQYKGU-UHFFFAOYSA-N
Synonym dichlorodiphenylmethane, diphenyldichloromethane, dpm halocarbon, benzophenone dichloride, alpha,alpha-dichlorodiphenylmethane, benzene, 1,1'-dichloromethylene bis, methane, dichlorodiphenyl, dichlorodiphenyl methane, dichloro phenyl methyl benzene, unii-81fez29ak6
PubChem CID 16327
IUPAC Name [dichloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(Cl)Cl

Specifications

Color Yellow
Boiling Point 193°C
Quantity 25 g
UN Number 1760
Formula Weight 237.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha,alpha-Dichlorodiphenylmethane