missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dichlorodiphenylmethane, 97%

Catalog No. AAL2044514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2044514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2044514 Supplier Thermo Scientific Chemicals Supplier No. L2044514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2051-90-3 | C13H10Cl2 | 237.123 g/mol

α,α-Dichlorodiphenylmethane is a degradation product of of polychloroaromatic insecticides such as Dicofol (D436700) and DDT. It is also used as a catalyst for photodegradation of plastics. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α,α-Dichlorodiphenylmethane is a degradation product of of polychloroaromatic insecticides such as Dicofol (D436700) and DDT. It is also used as a catalyst for photodegradation of plastics. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in water (reacts), ether, benzene, ethanol, and toluene.

Notes
Stable under recommended storage conditions. Incompatible with strong oxidizing agents and bases.

Chemical Identifiers

CAS 2051-90-3
Molecular Formula C13H10Cl2
Molecular Weight (g/mol) 237.123
MDL Number MFCD00000811
InChI Key OPTDDWCXQQYKGU-UHFFFAOYSA-N
Synonym dichlorodiphenylmethane, diphenyldichloromethane, dpm halocarbon, benzophenone dichloride, alpha,alpha-dichlorodiphenylmethane, benzene, 1,1'-dichloromethylene bis, methane, dichlorodiphenyl, dichlorodiphenyl methane, dichloro phenyl methyl benzene, unii-81fez29ak6
PubChem CID 16327
IUPAC Name [dichloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(Cl)Cl

Specifications

Density 1.242
Boiling Point 180°C to 181°C (17 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.605
Quantity 25 g
UN Number UN3265
Solubility Information Soluble in water (reacts),ether,benzene,ethanol,and toluene.
Formula Weight 237.13
Percent Purity 97%
Chemical Name or Material alpha,alpha-Dichlorodiphenylmethane
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 218-134-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only