missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,alpha'-Dichloro-o-xylene 99.0+%, TCI America™
$87.60 - $1004.73
Chemical Identifiers
| CAS | 612-12-4 |
|---|---|
| Molecular Formula | C8H8Cl2 |
| Molecular Weight (g/mol) | 175.05 |
| MDL Number | MFCD00000903 |
| InChI Key | FMGGHNGKHRCJLL-UHFFFAOYSA-N |
| Synonym | 1,2-bis chloromethyl benzene, o-xylylene dichloride, alpha,alpha'-dichloro-o-xylene, o-bis chloromethyl benzene, benzene, 1,2-bis chloromethyl, ccris 1773, o-xylene, alpha,alpha'-dichloro, benzene, 1,2-dichloromethyl, .alpha.,.alpha.'-dichloro-o-xylene, o-xylylenchlorid |
| PubChem CID | 11919 |
| IUPAC Name | 1,2-bis(chloromethyl)benzene |
| SMILES | ClCC1=CC=CC=C1CCl |
Chemical Identifiers
| 612-12-4 | |
| 175.05 | |
| FMGGHNGKHRCJLL-UHFFFAOYSA-N | |
| 11919 | |
| ClCC1=CC=CC=C1CCl |
| C8H8Cl2 | |
| MFCD00000903 | |
| 1,2-bis chloromethyl benzene, o-xylylene dichloride, alpha,alpha'-dichloro-o-xylene, o-bis chloromethyl benzene, benzene, 1,2-bis chloromethyl, ccris 1773, o-xylene, alpha,alpha'-dichloro, benzene, 1,2-dichloromethyl, .alpha.,.alpha.'-dichloro-o-xylene, o-xylylenchlorid | |
| 1,2-bis(chloromethyl)benzene |
Specifications
| 612-12-4 | |
| White-Yellow | |
| C8H8Cl2 | |
| 25 g | |
| 1,2-bis chloromethyl benzene, o-xylylene dichloride, alpha,alpha'-dichloro-o-xylene, o-bis chloromethyl benzene, benzene, 1,2-bis chloromethyl, ccris 1773, o-xylene, alpha,alpha'-dichloro, benzene, 1,2-dichloromethyl, .alpha.,.alpha.'-dichloro-o-xylene, o-xylylenchlorid | |
| ClCC1=CC=CC=C1CCl | |
| 175.05 | |
| 175.05 | |
| Crystalline Powder |
| 55°C | |
| 241°C | |
| MFCD00000903 | |
| 2928 | |
| FMGGHNGKHRCJLL-UHFFFAOYSA-N | |
| 1,2-bis(chloromethyl)benzene | |
| 11919 | |
| ≥99.0% (GC) | |
| alpha,alpha′-Dichloro-o-xylene |
Safety and Handling
EINECSNumber : (3)-0047
RTECSNumber : ZE4085000
TSCA : Yes