missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dichloro-o-xylene 99.0+%, TCI America™

Catalog No. D0429500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0429500G 500 g
D042925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0429500G Supplier TCI America Supplier No. D0429500G

Chemical Identifiers

CAS 612-12-4
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.05
MDL Number MFCD00000903
InChI Key FMGGHNGKHRCJLL-UHFFFAOYSA-N
Synonym 1,2-bis chloromethyl benzene, o-xylylene dichloride, alpha,alpha'-dichloro-o-xylene, o-bis chloromethyl benzene, benzene, 1,2-bis chloromethyl, ccris 1773, o-xylene, alpha,alpha'-dichloro, benzene, 1,2-dichloromethyl, .alpha.,.alpha.'-dichloro-o-xylene, o-xylylenchlorid
PubChem CID 11919
IUPAC Name 1,2-bis(chloromethyl)benzene
SMILES ClCC1=CC=CC=C1CCl

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 241°C
Quantity 500 g
UN Number 2928
Formula Weight 175.05
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Dichloro-o-xylene
EINECSNumber (3)-0047
RTECSNumber ZE4085000
TSCA Yes