missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,alpha'-Dibromo-m-xylene 97.0+%, TCI America™
$93.21 - $372.81
Chemical Identifiers
| CAS | 626-15-3 |
|---|---|
| Molecular Formula | C8H8Br2 |
| Molecular Weight (g/mol) | 263.96 |
| MDL Number | MFCD00000178 |
| InChI Key | OXHOPZLBSSTTBU-UHFFFAOYSA-N |
| Synonym | 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene |
| PubChem CID | 69373 |
| IUPAC Name | 1,3-bis(bromomethyl)benzene |
| SMILES | C1=CC(=CC(=C1)CBr)CBr |
Chemical Identifiers
| 626-15-3 | |
| 263.96 | |
| OXHOPZLBSSTTBU-UHFFFAOYSA-N | |
| 69373 | |
| C1=CC(=CC(=C1)CBr)CBr |
| C8H8Br2 | |
| MFCD00000178 | |
| 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene | |
| 1,3-bis(bromomethyl)benzene |
Specifications
| 626-15-3 | |
| White-Yellow | |
| C8H8Br2 | |
| 5 g | |
| 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene | |
| C1=CC(=CC(=C1)CBr)CBr | |
| 263.96 | |
| 263.96 | |
| Crystalline Powder |
| 76°C | |
| 140°C | |
| MFCD00000178 | |
| 2928 | |
| OXHOPZLBSSTTBU-UHFFFAOYSA-N | |
| 1,3-bis(bromomethyl)benzene | |
| 69373 | |
| ≥97.0% (GC) | |
| alpha,alpha′-Dibromo-m-xylene |
Safety and Handling
TSCA : Yes