missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha'-Tris(4-hydroxyphenyl)-1-ethyl-4-isopropylbenzene 98.0+%, TCI America™

Catalog No. T142825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T142825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T142825G Supplier TCI America Supplier No. T142825G

Chemical Identifiers

CAS 110726-28-8
Molecular Formula C29H28O3
Molecular Weight (g/mol) 424.54
MDL Number MFCD00191685
InChI Key WXYSZTISEJBRHW-UHFFFAOYSA-N
Synonym 4-[4-[1,1-Bis(4-hydroxyphenyl)ethyl]]-alpha,alpha-dimethylbenzylphenol
PubChem CID 634278
IUPAC Name 4-[2-[4-[1,1-bis(4-hydroxyphenyl)ethyl]phenyl]propan-2-yl]phenol
SMILES CC(C)(C1=CC=C(C=C1)C(C)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O)C4=CC=C(C=C4)O

Specifications

Melting Point 225°C
Color White
Quantity 25 g
Formula Weight 424.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha,alpha′-Tris(4-hydroxyphenyl)-1-ethyl-4-isopropylbenzene
EINECSNumber (4)-1644
TSCA Yes