Learn More
alpha,alpha,alpha-Trifluorotoluene, 99+%
CAS: 98-08-8 | C7H5F3 | 146.11 g/mol
$68.38 - $391.55
Chemical Identifiers
| CAS | 98-08-8 |
|---|---|
| Molecular Formula | C7H5F3 |
| Molecular Weight (g/mol) | 146.11 |
| MDL Number | MFCD00000372 |
| InChI Key | GETTZEONDQJALK-UHFFFAOYSA-N |
| Synonym | benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene |
| PubChem CID | 7368 |
| ChEBI | CHEBI:36810 |
| IUPAC Name | trifluoromethylbenzene |
| SMILES | C1=CC=C(C=C1)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC139770500
|
Thermo Scientific Chemicals
139770500 |
50 mL | Plastic bottle |
Each for $68.38
|
|
||||
|
AC139775000
|
Thermo Scientific Chemicals
139775000 |
500 mL | Plastic bottle |
Each for $136.54
|
|
||||
|
AC139770025
|
Thermo Scientific Chemicals
139770025 |
2.5 L | Plastic bottle |
Each for $391.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 98-08-8 | |
| 146.11 | |
| GETTZEONDQJALK-UHFFFAOYSA-N | |
| 7368 | |
| trifluoromethylbenzene |
| C7H5F3 | |
| MFCD00000372 | |
| benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene | |
| CHEBI:36810 | |
| C1=CC=C(C=C1)C(F)(F)F |
Specifications
| 98-08-8 | |
| 100.0 | |
| Colorless | |
| 102°C | |
| Authentic | |
| Plastic bottle | |
| 1.4130 to 1.415 | |
| MFCD00000372 | |
| 05, 290 | |
| 15, 1113 | |
| Solubility in water: may decompose. Other solubilities: soluble in many organic solvents | |
| C1=CC=C(C=C1)C(F)(F)F | |
| 146.11 | |
| CHEBI:36810 | |
| 99+% | |
| α, α, α-Trifluorotoluene, 99+% |
| 99.0 | |
| -29°C | |
| 1.1900g/mL | |
| 12°C | |
| 99% min. (GC) | |
| C7H5F3 | |
| C6H5CF3 | |
| 50 mL | |
| 1.19 | |
| benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene | |
| GETTZEONDQJALK-UHFFFAOYSA-N | |
| trifluoromethylbenzene | |
| 7368 | |
| 146.11 | |
| Liquid |
Safety and Handling
GHS H Statement:
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-635-0
RTECSNumber : XT9450000
TSCA : TSCA
RUO – Research Use Only