missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene 96.0+%, TCI America™

Catalog No. T005225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T005225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T005225G Supplier TCI America Supplier No. T005225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13209-15-9
Molecular Formula C8H6Br4
Molecular Weight (g/mol) 421.752
MDL Number MFCD00000131
InChI Key LNAOKZKISWEZNY-UHFFFAOYSA-N
Synonym 1,2-bis dibromomethyl benzene, alpha,alpha,alpha',alpha'-tetrabromo-o-xylene, benzene, 1,2-bis dibromomethyl, unii-3wg2h68vx4, o-xylylene tetrabromide, alpha,alpha,alpha,alpha-tetrabromo-o-xylene, a,a,a',a'-tetrabromo-o-xylene, o-xylene, alpha,alpha,alpha',alpha'-tetrabromo, alpha,alpha,beta,beta-tetrabromo-o-xylene, acmc-209boi
PubChem CID 83234
IUPAC Name 1,2-bis(dibromomethyl)benzene
SMILES C1=CC=C(C(=C1)C(Br)Br)C(Br)Br

Specifications

Melting Point 115°C
Quantity 25 g
UN Number 1759
Formula Weight 421.75
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha,alpha′,alpha′-Tetrabromo-o-xylene