missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%

Catalog No. p-7021688
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1412118 50 g
AAA1412130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1412118 Supplier Thermo Scientific Chemicals Supplier No. A1412118
Only null left

CAS: 13209-15-9 | C8H6Br4 | 421.752 g/mol

α,α,α',α'-Tetrabromo-o-xylene reacts with 5endo,6endo-bis-chlormethyl-norborn-2-en to obtain trans/cis-2,3-bis(chloromethyl)-1,4-methano-1,2,3,4-tetrahydroanthracene. Further, it is involved in the preparation of 1-(3-butenyl)-2-(deuteriomethyl)benzene by reacting with allylmagnesium bromide. In addition this, it is used to prepare exo-benzocyclobuteno-1,2,3,4-tetrahydro-1,4-methanoanthracene by reacting with exo-1,4,4a,8b-tetrahydro-1,4-methanobiphenylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α,α,α′,α′-Tetrabromo-o-xylene reacts with 5endo,6endo-bis-chlormethyl-norborn-2-en to obtain trans/cis-2,3-bis(chloromethyl)-1,4-methano-1,2,3,4-tetrahydroanthracene. Further, it is involved in the preparation of 1-(3-butenyl)-2-(deuteriomethyl)benzene by reacting with allylmagnesium bromide. In addition this, it is used to prepare exo-benzocyclobuteno-1,2,3,4-tetrahydro-1,4-methanoanthracene by reacting with exo-1,4,4a,8b-tetrahydro-1,4-methanobiphenylene.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 13209-15-9
Molecular Formula C8H6Br4
Molecular Weight (g/mol) 421.752
MDL Number MFCD00000131
InChI Key LNAOKZKISWEZNY-UHFFFAOYSA-N
Synonym 1,2-bis dibromomethyl benzene, alpha,alpha,alpha',alpha'-tetrabromo-o-xylene, benzene, 1,2-bis dibromomethyl, unii-3wg2h68vx4, o-xylylene tetrabromide, alpha,alpha,alpha,alpha-tetrabromo-o-xylene, a,a,a',a'-tetrabromo-o-xylene, o-xylene, alpha,alpha,alpha',alpha'-tetrabromo, alpha,alpha,beta,beta-tetrabromo-o-xylene, acmc-209boi
PubChem CID 83234
IUPAC Name 1,2-bis(dibromomethyl)benzene
SMILES C1=CC=C(C(=C1)C(Br)Br)C(Br)Br

Specifications

Melting Point 114°C to 118°C
Quantity 50 g
UN Number UN1759
Beilstein 2211698
Solubility Information Insoluble in water.
Formula Weight 421.75
Percent Purity 97%
Chemical Name or Material alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H317-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 236-176-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only