missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha',2,3,5,6-Hexachloro-p-xylene 98.0+%, TCI America™

Catalog No. H00655G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H00655G Supplier TCI America Supplier No. H00655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1079-17-0
Molecular Formula C8H4Cl6
Molecular Weight (g/mol) 312.82
MDL Number MFCD00000894
InChI Key IYGDLOMSJZQSGY-UHFFFAOYSA-N
PubChem CID 66179
IUPAC Name 1,2,4,5-tetrachloro-3,6-bis(chloromethyl)benzene
SMILES C(C1=C(C(=C(C(=C1Cl)Cl)CCl)Cl)Cl)Cl

Specifications

Melting Point 181°C
Quantity 5 g
UN Number 3261
Formula Weight 312.82
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′,2,3,5,6-Hexachloro-p-xylene