missing translation for 'onlineSavingsMsg'
Learn More

alpha-(4-Piperidyl)benzhydrol 99.0+%, TCI America™

Catalog No. P13695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13695G 5 g
P136925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13695G Supplier TCI America Supplier No. P13695G

Chemical Identifiers

CAS 115-46-8
Molecular Formula C18H21NO
Molecular Weight (g/mol) 267.372
MDL Number MFCD00066980
InChI Key ZMISODWVFHHWNR-UHFFFAOYSA-N
Synonym azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol
PubChem CID 15723
IUPAC Name diphenyl(piperidin-4-yl)methanol
SMILES C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 163°C
Color White
Quantity 5 g
Formula Weight 267.37
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-(4-Piperidyl)benzhydrol
RTECSNumber TN0470000
TSCA No