missing translation for 'onlineSavingsMsg'
Learn More

alpha-2-Chloroisodurene, 98%, Thermo Scientific™

$58.24 - $58.24

Chemical Identifiers

CAS 1585-16-6
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.66
MDL Number MFCD00000901
InChI Key UNRGEIXQCZHICP-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene
PubChem CID 74108
IUPAC Name 2-(chloromethyl)-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(CCl)C(C)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC159780050
SDS View Documents
Thermo Scientific Chemicals
159780050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC159780250
SDS View Documents
Thermo Scientific Chemicals
159780250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1585-16-6
168.66
UNRGEIXQCZHICP-UHFFFAOYSA-N
74108
CC1=CC(C)=C(CCl)C(C)=C1
C10H13Cl
MFCD00000901
2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene
2-(chloromethyl)-1,3,5-trimethylbenzene

Specifications

1585-16-6
100.0
106°C
97.5% min. (GC)
C10H13Cl
MFCD00000901
2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene
UNRGEIXQCZHICP-UHFFFAOYSA-N
2-(chloromethyl)-1,3,5-trimethylbenzene
74108
98%
97.5
114°C to 115°C (10.0mmHg)
Authentic
Glass bottle
(CH3)3C6H2CH2Cl
5g
Solubility in water: insoluble
CC1=CC(C)=C(CCl)C(C)=C1
168.66
168.67
α-2-Chloroisodurene, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep only in

GHS Signal Word: Danger

EINECSNumber : 216-440-3

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only