missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha-1D Adrenoceptor Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572171
Description
alpha-1D Adrenoceptor Polyclonal Antibody for Western Blot, IHC (P), Flow
Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1D-adrenergic receptor. Similar to alpha-1B-adrenergic receptor gene, this gene comprises 2 exons and a single intron that interrupts the coding region.Chemical Identifiers
| alpha-1D Adrenoceptor | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| Adra1d | |
| [a]1d; ADA1D; Adra1; Adra-1; Adra1a; Adra1d; ADRA1R; Adrd1; adrenergic receptor delta1; adrenergic receptor, alpha 1d; adrenergic, alpha -1D-, receptor; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1D-, receptor; adrenoceptor alpha 1D; alpha 1D-adrenoceptor; alpha 1D-adrenoreceptor; ALPHA1; alpha-1A adrenergic receptor; Alpha-1D adrenergic receptor; alpha-1D adrenoceptor; Alpha-1D adrenoreceptor; alpha-1D-adrenergic receptor; alpha1D-AR; Alpha1delta-AR; alpha-adrenergic receptor 1a; DAR; dJ779E11.2; Gpcr8; RA42; Spr8 | |
| Adra1d | |
| Primary | |
| Antigen Affinity Chromatography, Protein A |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P23944, P25100 | |
| 146, 29413 | |
| KLH conjugated synthetic peptide between 1-30amino acids from the N-terminal region of human ADRA1D. | |
| Antibody |
Specifications
| alpha-1D Adrenoceptor | |
| Polyclonal | |
| Unconjugated | |
| Adra1d | |
| [a]1d; ADA1D; Adra1; Adra-1; Adra1a; Adra1d; ADRA1R; Adrd1; adrenergic receptor delta1; adrenergic receptor, alpha 1d; adrenergic, alpha -1D-, receptor; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1D-, receptor; adrenoceptor alpha 1D; alpha 1D-adrenoceptor; alpha 1D-adrenoreceptor; ALPHA1; alpha-1A adrenergic receptor; Alpha-1D adrenergic receptor; alpha-1D adrenoceptor; Alpha-1D adrenoreceptor; alpha-1D-adrenergic receptor; alpha1D-AR; Alpha1delta-AR; alpha-adrenergic receptor 1a; DAR; dJ779E11.2; Gpcr8; RA42; Spr8 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 146, 29413 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P23944, P25100 | |
| Adra1d | |
| KLH conjugated synthetic peptide between 1-30amino acids from the N-terminal region of human ADRA1D. | |
| 400 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |