missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1 Antitrypsin Monoclonal Antibody (1AT11-210.1), Invitrogen™
Mouse Monoclonal Antibody
$746.91 - $1850.60
Specifications
| Antigen | alpha-1 Antitrypsin |
|---|---|
| Clone | 1AT11-210.1 |
| Concentration | 1.2 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
PIMA546034
|
Invitrogen™
MA546034 |
100 μg |
Each for $746.91
|
|
|||||
|
01-671-579
|
Invitrogen™
MA110626 |
500 μg |
Each for $859.85
|
|
|||||
|
PIMA546019
|
Invitrogen™
MA546019 |
1 mg |
Each for $1,850.60
|
|
|||||
Description
MA1-10626 detects alpha-1 Antitrypsin from human samples. MA1-10626 does not cross react with alpha-1-antichymotrysin. MA1-10626 has been successfully used in ELISA applications. The MA1-10626 immunogen is purified human serum alpha-1 Antitrypsin.
Alpha-1-AT is synthesized in the liver and it acts as an inhibitor of proteases such as trypsin, elastase, chymotrypsin, collagenase, leucocytic proteases, plasmin, and thrombin, which may be released during inflammatory reactions in the lung. In the absence of alpha-1-AT, these enzymes are not inhibited and they may digest pulmonary parenchyma. Alpha-1-AT deficiency is associated with chronic obstructive lung disease (emphysema) and less frequently with hepatic cirrhosis in infants and respiratory distress of the newborn. Increase in alpha-1-AT occurs as an acute phase response to tissue necrosis and inflammation. Serum level of alpha-1-AT is elevated in rheumatoid arthritis, bacterial infections, vasculitis, and carcinomatosis.Specifications
| alpha-1 Antitrypsin | |
| 1.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| a-1 AT; A1A; a1-antitrypsin; A1AT; AAT; Aat2; Aat-2; alpha 1 antitrypsin; alpha-1 antitrypsin; alpha-1 protease inhibitor; Alpha-1 protease inhibitor 1; alpha-1-antiproteinase; Alpha-1-antitrypsin; alpha-1-antitrypsin (protease inhibitor); alpha-1-antitrypsin 1-1; alpha-1-antitrypsin 2; alpha-1-antitrypsin null; alpha1AT; alpha-1-protease inhibitor; alpha-1-proteinase inhibitor; Digestive Zymogen; Dom1; MGC23330; MGC9222; PI; PI1; PRO0684; PRO2209; PRO2275; protease inhibitor 1 (anti-elastase), alpha-1-antitrypsin; protease inhibitor 1 (anti-elastase, antitrypsin, alpha-1); Protease Serine 1; serine (or cysteine) peptidase inhibitor, clade A, member 1A; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A (alpha-1antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1a; Serine Protease 1; serine protease inhibitor 1-1; Serine protease inhibitor A1a; serine protease inhibitor alpha 1; serine proteinase inhibitor, clade A, member 1; serpin A1; Serpin A1a; serpin family A member 1; serpin peptidase inhibitor clade A member 1; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; SERPINA1; Serpina1a; Short peptide from AAT; SPAAT; Spi1; Spi1-1; Spi1-3; TRP1; TRY1 | |
| SERPINA1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1AT11-210.1 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01009 | |
| 5265 | |
| Purified human serum alpha-1 Antitrypsin. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| SERPINA1 |