missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ALP Monoclonal Antibody (K52014_16F8), Invitrogen™

Catalog No. MA555957
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555957 50 μL
MA556337 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555957 Supplier Invitrogen™ Supplier No. MA555957
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MNPIVVVHGG GAGPISKDRK ERVHQGMVRA ATVGYGILRE GGSAVDAVEG AVVALEDDPE FNAGCGSVLN TNGEVEMDAS IMDGKDLSAG AVSAVQCIAN PIKLARLVME KTPHCFLTDQ GAAQFAAAMG VPEIPGEKLV TERNKKRLEK EKHEKGAQKT DCQKNLGTVG AVALDCKGNV AYATSTGGIV NKMVGRVGDS PCLGAGGYAD NDIGAVSTTG HGESILKVNL ARLTLFHIEQ GKTVEEAADL SLGYMKSRVK GLGGLIVVSK TGDWVAKWTS TSMPWAAAKD GKLHFGIDPD DTTITDLP

ALP is found in a variety of tumors but only occasionally in normal tissues. Ovarian cancers are highly positive. ALP is further found in brain tumors, some colonic cancers, some lung cancers, some prostate cancers, uterine cancers, some breast cancers and some thyroid cancers. It was not found in serum.

Specifications

Antigen ALP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K52014_16F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Asrgl1
Gene Accession No. Q7L266
Gene Alias 2410004D18Rik; ALP; ALP1; asparaginase and isoaspartyl peptidase 1; asparaginase like 1; asparaginase-like 1; asparaginase-like 1 protein; asparaginase-like protein 1; Asparaginase-like sperm autoantigen; ASRGL1; AU040643; AW060375; Beta-aspartyl-peptidase; CRASH; FLJ22316; Glial asparaginase; Gliap; Hiob; Hiob2; isoaspartyl dipeptidase; Isoaspartyl peptidase/L-asparaginase; Isoaspartyl peptidase/L-asparaginase alpha chain; Isoaspartyl peptidase/L-asparaginase beta chain; L-asparaginase; L-asparagine amidohydrolase; testis secretory sperm-binding protein Li 242mP; zgc:103568
Gene Symbols Asrgl1
Host Species Mouse
Immunogen Recombinant human ASRGL1 (immunogen range is 1-308).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80150
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less