missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyltriphenyltin 95.0+%, TCI America™
Supplier: TCI America A14915G
Specifications
| Allyltriphenyltin | |
| 74°C | |
| C21H20Sn | |
| 5 g | |
| Triphenylallyltin | |
| C=CC[Sn](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 391.101 | |
| 391.10 | |
| Crystalline Powder |
| 76-63-1 | |
| White-Yellow | |
| MFCD00048162 | |
| 3146 | |
| NDUYAGLANMHJHF-UHFFFAOYSA-N | |
| triphenyl(prop-2-enyl)stannane | |
| 6452 | |
| ≥95.0% (W) |
Chemical Identifiers
| 76-63-1 | |
| 391.101 | |
| NDUYAGLANMHJHF-UHFFFAOYSA-N | |
| 6452 | |
| C=CC[Sn](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 |
| C21H20Sn | |
| MFCD00048162 | |
| Triphenylallyltin | |
| triphenyl(prop-2-enyl)stannane |
Safety and Handling
RTECSNumber : WH6705000
TSCA : No