missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenyltin 95.0+%, TCI America™

Catalog No. p-7117472
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14915G 5 g
A149125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14915G Supplier TCI America Supplier No. A14915G
Only null left

Chemical Identifiers

CAS 76-63-1
Molecular Formula C21H20Sn
Molecular Weight (g/mol) 391.101
MDL Number MFCD00048162
InChI Key NDUYAGLANMHJHF-UHFFFAOYSA-N
Synonym Triphenylallyltin
PubChem CID 6452
IUPAC Name triphenyl(prop-2-enyl)stannane
SMILES C=CC[Sn](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 5 g
UN Number 3146
Formula Weight 391.10
Percent Purity ≥95.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Allyltriphenyltin
RTECSNumber WH6705000
TSCA No