missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. A100725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A100725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A100725G Supplier TCI America Supplier No. A100725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1560-54-9
Molecular Formula C21H20BrP
Molecular Weight (g/mol) 383.27
MDL Number MFCD00011808
InChI Key FWYKRJUVEOBFGH-UHFFFAOYSA-M
Synonym allyltriphenylphosphonium bromide, triphenyl allylphosphonium bromide, phosphonium, triphenyl-2-propenyl-, bromide, allyl triphenylphosphonium bromide, phosphonium, allyltriphenyl-, bromide, allyltriphenylphosphonium bromide tal, triphenyl prop-2-enyl phosphanium bromide, 2-propenyltriphenylphosphonium bromide, triphenyl prop-2-en-1-yl phosphanium bromide, wln: 1u2pr&r&r &e
PubChem CID 197740
IUPAC Name triphenyl(prop-2-en-1-yl)phosphanium bromide
SMILES [Br-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 225°C
Color White
Quantity 25 g
Formula Weight 383.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Allyltriphenylphosphonium Bromide
RTECSNumber TA1843000
TSCA No