missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenylphosphonium bromide, 99%

Catalog No. AC103041000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103041000 100 g Glass bottle
AC103040250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103041000 Supplier Thermo Scientific Chemicals Supplier No. 103041000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1560-54-9 | C21H20BrP | 383.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1560-54-9
Molecular Formula C21H20BrP
Molecular Weight (g/mol) 383.27
MDL Number MFCD00011808
InChI Key FWYKRJUVEOBFGH-UHFFFAOYSA-M
Synonym allyltriphenylphosphonium bromide, triphenyl allylphosphonium bromide, phosphonium, triphenyl-2-propenyl-, bromide, allyl triphenylphosphonium bromide, phosphonium, allyltriphenyl-, bromide, allyltriphenylphosphonium bromide tal, triphenyl prop-2-enyl phosphanium bromide, 2-propenyltriphenylphosphonium bromide, triphenyl prop-2-en-1-yl phosphanium bromide, wln: 1u2pr&r&r &e
PubChem CID 197740
IUPAC Name triphenyl(prop-2-enyl)phosphanium;bromide
SMILES [Br-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 222.0°C to 225.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2C=CHCH2P(C6H5)3Br
Quantity 100 g
Solubility Information Solubility in water: decomposes
Formula Weight 383.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Allyltriphenylphosphonium bromide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-332-6
RTECSNumber TA1843000

RUO – Research Use Only