missing translation for 'onlineSavingsMsg'
Learn More

Allyltriphenylphosphonium bromide, 99%

Catalog No. AAA1449018
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1449018 50 g
AAA1449030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1449018 Supplier Thermo Scientific Chemicals Supplier No. A1449018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1560-54-9 | C21H20BrP | 383.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1560-54-9
Molecular Formula C21H20BrP
Molecular Weight (g/mol) 383.27
MDL Number MFCD00011808
InChI Key FWYKRJUVEOBFGH-UHFFFAOYSA-M
Synonym allyltriphenylphosphonium bromide, triphenyl allylphosphonium bromide, phosphonium, triphenyl-2-propenyl-, bromide, allyl triphenylphosphonium bromide, phosphonium, allyltriphenyl-, bromide, allyltriphenylphosphonium bromide tal, triphenyl prop-2-enyl phosphanium bromide, 2-propenyltriphenylphosphonium bromide, triphenyl prop-2-en-1-yl phosphanium bromide, wln: 1u2pr&r&r &e
PubChem CID 197740
IUPAC Name triphenyl(prop-2-enyl)phosphanium;bromide
SMILES [Br-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 222°C to 225°C
Linear Formula (CH2=CHCH2)(C6H5)3PBr
Quantity 50 g
Beilstein 3579053
Sensitivity Hygroscopic
Formula Weight 383.27
Percent Purity 99%
Chemical Name or Material Allyltriphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-332-6
RTECSNumber TA1843000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only