missing translation for 'onlineSavingsMsg'
Learn More

Allyltrimethylsilane, 99%

CAS: 762-72-1 | C6H14Si | 114.27 g/mol

$129.02 - $353.14

Chemical Identifiers

CAS 762-72-1
Molecular Formula C6H14Si
Molecular Weight (g/mol) 114.27
InChI Key HYWCXWRMUZYRPH-UHFFFAOYSA-N
Synonym allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane
PubChem CID 69808
IUPAC Name trimethyl(prop-2-enyl)silane
SMILES C[Si](C)(C)CC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC196990100
SDS View Documents
Thermo Scientific Chemicals
196990100
10 mL Glass bottle N/A
Only null left
 
AC196990500
SDS View Documents
Thermo Scientific Chemicals
196990500
50 mL Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

762-72-1
114.27
allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane
trimethyl(prop-2-enyl)silane
C6H14Si
HYWCXWRMUZYRPH-UHFFFAOYSA-N
69808
C[Si](C)(C)CC=C

Specifications

762-72-1
0.7190g/mL
7°C
99%
C6H14Si
H2C=CHCH2Si(CH3)3
10,6; 11,16; 12,23; 13,11; 14,18; 15,8
15,29
Solubility in water: insoluble
C[Si](C)(C)CC=C
114.27
114.27
Liquid
Colorless
85.0°C
Authentic
Glass bottle
1.4060 to 1.4080
10 mL
0.719
allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane
HYWCXWRMUZYRPH-UHFFFAOYSA-N
trimethyl(prop-2-enyl)silane
69808
99%
Allyltrimethylsilane
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 212-104-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only