missing translation for 'onlineSavingsMsg'
Learn More

Allyltrimethoxysilane 97.0+%, TCI America™

Catalog No. a15045ml
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15045ML 5 mL
A150425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15045ML Supplier TCI America Supplier No. A15045ML
Only null left

Chemical Identifiers

CAS 2551-83-9
Molecular Formula C6H14O3Si
Molecular Weight (g/mol) 162.26
MDL Number MFCD00053865
InChI Key LFRDHGNFBLIJIY-UHFFFAOYSA-N
Synonym allyltrimethoxysilane, silane, trimethoxy-2-propenyl, silane, trimethoxy-2-propen-1-yl, allyl trimethoxy silane, silane, allyltrimethoxy, allytrimethoxysilane, acmc-209gkn, trimethoxy prop-2-enyl silane, silane,trimethoxy-2-propen-1-yl
PubChem CID 75698
IUPAC Name trimethoxy(prop-2-enyl)silane
SMILES CO[Si](CC=C)(OC)OC

Specifications

Color Yellow
Boiling Point 136°C
Quantity 5 mL
UN Number 1993
Formula Weight 162.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyltrimethoxysilane
EINECSNumber (2)-2066
TSCA Yes