missing translation for 'onlineSavingsMsg'
Learn More

Allyltriethoxysilane 96.0+%, TCI America™

Catalog No. A078510ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A078510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A078510ML Supplier TCI America Supplier No. A078510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2550-04-1
Molecular Formula C9H20O3Si
Molecular Weight (g/mol) 204.34
MDL Number MFCD00009064
InChI Key UMFJXASDGBJDEB-UHFFFAOYSA-N
Synonym allyltriethoxysilane, silane, triethoxy-2-propenyl, triethoxyallylsilane, silane, allyltriethoxy, allytriethoxysilane, triethoxy prop-2-en-1-yl silane, silane, triethoxy-2-propen-1-yl, allyl triethoxysilane, acmc-2097bb
PubChem CID 75692
IUPAC Name triethoxy(prop-2-enyl)silane
SMILES CCO[Si](CC=C)(OCC)OCC

Specifications

Color Yellow
Boiling Point 176°C
Quantity 10 mL
UN Number 1993
Formula Weight 204.34
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyltriethoxysilane
EINECSNumber (2)-2066
TSCA Yes