accessibility menu, dialog, popup

UserName

CAS RN 2550-04-1

IUPAC Name: triethoxy(prop-2-en-1-yl)silane
Synonyms: triethoxy(prop-2-en-1-yl)silane
Molecular Weight (g/mol): 204.34
Molecular Formula: C9H20O3Si
InChi Key: UMFJXASDGBJDEB-UHFFFAOYSA-N
SMILES: CCO[Si](CC=C)(OCC)OCC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)