missing translation for 'onlineSavingsMsg'
Learn More

Allyltriethoxysilane, 97%

Catalog No. p-7044057
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0429706 5 g
AAL0429714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0429706 Supplier Thermo Scientific Chemicals Supplier No. L0429706

CAS: 2550-04-1 | C9H20O3Si | 204.34 g/mol

Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents. Incompatible with moisture and oxidizing agents.

Chemical Identifiers

CAS 2550-04-1
MDL Number MFCD00009064

Specifications

Boiling Point 176°C to 177°C
Quantity 5 g
Molecular Formula C9H20O3Si
Linear Formula CH2=CHCH2Si(OCH2CH3)3
UN Number UN1993
Beilstein 1772306
Flash Point 21°C (69°F)
Synonym allyltriethoxysilane, silane, triethoxy-2-propenyl, triethoxyallylsilane, silane, allyltriethoxy, allytriethoxysilane, triethoxy prop-2-en-1-yl silane, silane, triethoxy-2-propen-1-yl, allyl triethoxysilane, acmc-2097bb
Solubility Information Hydrolyzes in water.
InChI Key UMFJXASDGBJDEB-UHFFFAOYSA-N
SMILES CCO[Si](CC=C)(OCC)OCC
IUPAC Name triethoxy(prop-2-enyl)silane
Molecular Weight (g/mol) 204.34
PubChem CID 75692
Formula Weight 204.34
Percent Purity 97%
Refractive Index 1.407
Sensitivity Moisture sensitive
Density 0.953
Chemical Name or Material Allyltriethoxysilane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 219-843-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only