missing translation for 'onlineSavingsMsg'
Learn More

Allylthiourea, 98%

Catalog No. p-52280
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
AC148800250 25 g
AC148801000 100 g
AC148805000 500 g
3 options

Catalog No. AC148800250

Supplier: Thermo Scientific Chemicals 148800250

Only null left

CAS: 109-57-9 | C4H8N2S | 116.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-57-9
MDL Number MFCD00004940
InChI Key HTKFORQRBXIQHD-UHFFFAOYSA-N
Synonym allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
PubChem CID 1549517
ChEBI CHEBI:74079
IUPAC Name prop-2-enylthiourea
SMILES C=CCNC(=S)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70.0°C to 78.0°C
Color White to Beige
Loss on Drying 1.5% max. (50°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H8N2S
Linear Formula H2C=CHCH2NHCSNH2
Quantity 25 g
Beilstein 04, 211
Merck Index 15, 9515
Solubility Information Solubility in water: 67g/L (20°C). Other solubilities: 1370g/L methanol (20°C)- insoluble in benzene,soluble in ether,solutions of borax,benzoates,and urethane,slightly soluble in 70% alcohol
Molecular Weight (g/mol) 116.18
Formula Weight 116.18
Percent Purity 98%
Physical Form Crystals, Powder, Granules or Chunks
Chemical Name or Material Allylthiourea, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 203-683-5
RTECSNumber YR8050000
TSCA TSCA

RUO – Research Use Only