Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylsuccinic Anhydride 97.0+%, TCI America™

Catalog No. p-7117348
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12835G 5 g
A128325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. A12835G missing translation for 'mfr' TCI America missing translation for 'supplierNo' A12835G
Only null left

Chemical Identifiers

CAS 7539-12-0
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00085227
InChI Key WUMMIJWEUDHZCL-UHFFFAOYSA-N
Synonym 4-Pentene-1,2-dicarboxylic Anhydride, (2-Propenyl)butanedioic Anhydride
PubChem CID 111009
IUPAC Name 3-prop-2-enyloxolane-2,5-dione
SMILES C=CCC1CC(=O)OC1=O

Specifications

Color Yellow
Boiling Point 143°C
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2810
Formula Weight 140.14
Percent Purity ≥97.0% (GC)
Chemical Name or Material Allylsuccinic Anhydride
EINECSNumber (5)-7013
RTECSNumber WN1050000
TSCA No