accessibility menu, dialog, popup

UserName

CH2OOO

CAS RN 7539-12-0

IUPAC Name: 3-(prop-2-en-1-yl)oxolane-2,5-dione
Synonyms: succinic anhydride, allyl-
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: WUMMIJWEUDHZCL-UHFFFAOYNA-N
SMILES: C=CCC1CC(=O)OC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results