missing translation for 'onlineSavingsMsg'
Learn More

Allylpalladium(II) Chloride Dimer 97.0+%, TCI America™

$322.48 - $643.10

Chemical Identifiers

CAS 12012-95-2
Molecular Formula C6H10Cl2Pd2
Molecular Weight (g/mol) 365.89
MDL Number MFCD00044874
InChI Key PENAXHPKEVTBLF-UHFFFAOYSA-L
Synonym allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
PubChem CID 61538
IUPAC Name Allylpalladium chloride dimer
SMILES Cl[Pd].Cl[Pd].c:c:c.c:c:c
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A1479500MG
View Documents
TCI America
A1479500MG
500 mg
Each for $322.48
Only null left
 
A14791G
View Documents
TCI America
A14791G
1 g
Each for $643.10
Only null left
 
Description

Description

Specifications

Chemical Identifiers

12012-95-2
365.89
PENAXHPKEVTBLF-UHFFFAOYSA-L
61538
Cl[Pd].Cl[Pd].c:c:c.c:c:c
C6H10Cl2Pd2
MFCD00044874
allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
Allylpalladium chloride dimer

Specifications

12012-95-2
MFCD00044874
allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
Cl[Pd].Cl[Pd].c:c:c.c:c:c
365.89
365.89
Crystalline Powder
C6H10Cl2Pd2
500 mg
PENAXHPKEVTBLF-UHFFFAOYSA-L
Allylpalladium chloride dimer
61538
≥97.0% (T)
Allylpalladium(II) Chloride Dimer
Videos
Safety and Handling

Safety and Handling

RTECSNumber : RT3510000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'