missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allylpalladium(II) Chloride Dimer 97.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 365.89 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Allylpalladium(II) Chloride Dimer |
Safety and Handling
| RTECSNumber | RT3510000 |
| TSCA | Yes |