missing translation for 'onlineSavingsMsg'
Learn More

Allylpalladium(II) chloride dimer, Pd 56.0% min

Catalog No. p-7048276
Change view
Click to view available options
Quantity:
0.25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1000501 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1000501 Supplier Thermo Scientific Chemicals Supplier No. 01000501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12012-95-2 | C6H10Cl2Pd2 | 365.89 g/mol

Allylpalladium(II) chloride dimer is used as an important catalyst in Heck reaction. It reacts with cyclopentadienyl anion to give cyclopentadienyl allyl palladium. It acts as a precatalyst for asymmetric and cross-coupling catalysis. Further, it is used in the preparation of 1,4-diallyl-1,2-dihydroisoquinolines. It is also employed as a catalyst for greener Buchwald-Hartwig coupling reaction and involved in the synthesis of cationic palladium catalysts. In addition, it takes part in preparation of -heterocyclic carbene-palladium-eta3-allyl chloride complex, which is an efficient catalyst for the Suzuki-Miyaura cross-coupling reactions in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12012-95-2
Molecular Formula C6H10Cl2Pd2
Molecular Weight (g/mol) 365.89
MDL Number MFCD00044874
InChI Key PENAXHPKEVTBLF-UHFFFAOYSA-L
Synonym allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
PubChem CID 61538
IUPAC Name chloropalladium(1+);prop-1-ene
SMILES Cl[Pd].Cl[Pd].c:c:c.c:c:c

Specifications

Melting Point 120°C (decomposition)
Color Yellow
Quantity 0.25 g
Beilstein 4124623
Sensitivity Air and moisture sensitive
Solubility Information Very soluble in dichloromethane and dichloroethane; Slightly soluble in toluene,insoluble in waterSoluble in chloroform,benzene,acetone and dichloromethane,dichloroethane and methanol; Slightly soluble in toluene; Insoluble in water.
Formula Weight 365.85
Concentration or Composition (by Analyte or Components) Pd 56.0% min.
Physical Form Powder
Chemical Name or Material Allylpalladium(II) chloride dimer
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 234-579-8
RTECSNumber RT3510000
TSCA No
storageNote1 2°C to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only