missing translation for 'onlineSavingsMsg'
Learn More

Allylcyclopentane, 97%

Catalog No. p-7024953
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2022806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2022806 Supplier Thermo Scientific Chemicals Supplier No. B2022806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3524-75-2 | C8H14 | 110.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3524-75-2
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00001390
InChI Key NHIDGVQVYHCGEK-UHFFFAOYSA-N
Synonym allylcyclopentane, 3-cyclopentylpropene, cyclopentane, 2-propenyl, cyclopentane, allyl, 3-cyclopentyl-1-propene, allyl cyclopentane, 2-propenylcyclopentane, 1-propene, 3-cyclopentyl, 3-cyclopentylprop-1-ene
PubChem CID 77059
IUPAC Name prop-2-enylcyclopentane
SMILES C=CCC1CCCC1

Specifications

Melting Point -111°C
Density 0.793
Boiling Point 125°C to 127°C
Flash Point 12°C (53°F)
Refractive Index 1.441
Quantity 5 g
UN Number UN3295
Beilstein 2036419
Formula Weight 110.2
Percent Purity 98%
Chemical Name or Material Allylcyclopentane
Show More Show Less
Hazard Category H225-H302+H332
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 222-542-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only