accessibility menu, dialog, popup

UserName

CAS RN 3524-75-2

IUPAC Name: (prop-2-en-1-yl)cyclopentane
Synonyms: allylcyclopentane
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: NHIDGVQVYHCGEK-UHFFFAOYSA-N
SMILES: C=CCC1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results