missing translation for 'onlineSavingsMsg'
Learn More

Allylboronic acid pinacol ester, 98%

Catalog No. AC395130010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395130010 1 g Glass bottle
AC395130050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC395130010 Supplier Thermo Scientific Chemicals Supplier No. 395130010

CAS: 72824-04-5 | C9H17BO2 | 168.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72824-04-5
Molecular Formula C9H17BO2
Molecular Weight (g/mol) 168.04
MDL Number MFCD00013347
InChI Key YMHIEPNFCBNQQU-UHFFFAOYSA-N
Synonym allylboronic acid pinacol ester, 2-allyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, allylboronic acid, pinacol cyclic ester, 4,4,5,5-tetramethyl-2-prop-2-en-1-yl-1,3,2-dioxaborolane, allylboronic acid piracol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-propenyl, 2-prop-2-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, allylboronicacidpinacolester, pinacol allylboronate, abape
PubChem CID 2763171
IUPAC Name 4,4,5,5-tetramethyl-2-prop-2-enyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(CC=C)OC1(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8900g/mL
Boiling Point 50.0°C to 53.0°C (5.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble
Formula Weight 168.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allylboronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only