missing translation for 'onlineSavingsMsg'
Learn More

Allylbenzene 98.0+%, TCI America™

Catalog No. p-7134695
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P060925ML 25 mL
P0609100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P060925ML Supplier TCI America Supplier No. P060925ML
Only null left

Chemical Identifiers

CAS 300-57-2
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00008651
InChI Key HJWLCRVIBGQPNF-UHFFFAOYSA-N
Synonym allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene
PubChem CID 9309
IUPAC Name prop-2-enylbenzene
SMILES C=CCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 mL
UN Number 3295
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allylbenzene
RTECSNumber CY2275000
TSCA Yes