missing translation for 'onlineSavingsMsg'
Learn More

Allylbenzene, 98%

Catalog No. AAA1296522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1296522 100 g
AAA1296514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1296522 Supplier Thermo Scientific Chemicals Supplier No. A1296522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 300-57-2 | C9H10 | 118.179 g/mol

Allylbenzene is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylbenzene is used in organic synthesis.

Solubility
Insoluble in water. Soluble in alcohol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 300-57-2
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00008651
InChI Key HJWLCRVIBGQPNF-UHFFFAOYSA-N
Synonym allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene
PubChem CID 9309
IUPAC Name prop-2-enylbenzene
SMILES C=CCC1=CC=CC=C1

Specifications

Melting Point ∼-50°C
Density 0.892
Boiling Point 158°C to 160°C
Flash Point 33°C (91°F)
Odor Aromatic
Refractive Index 1.51
Quantity 100 g
UN Number UN3295
Beilstein 1098501
Solubility Information Insoluble in water. Soluble in alcohol,ether and benzene.
Formula Weight 118.18
Percent Purity 98%
Chemical Name or Material Allylbenzene
Show More Show Less
Hazard Category H226-H304
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 206-095-7
RTECSNumber CY2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only