missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allylamine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America A0226500G
Specifications
| Allylamine Hydrochloride | |
| 106°C | |
| MFCD00054329 | |
| 2811 | |
| MLGWTHRHHANFCC-UHFFFAOYSA-N | |
| hydrogen prop-2-en-1-amine chloride | |
| 82291 | |
| ≥98.0% (T) |
| 10017-11-5 | |
| C3H8ClN | |
| 500 g | |
| 3-Aminopropene Hydrochloride | |
| [H+].[Cl-].NCC=C | |
| 93.55 | |
| 93.55 | |
| Crystalline Powder |
Chemical Identifiers
| 10017-11-5 | |
| 93.55 | |
| MLGWTHRHHANFCC-UHFFFAOYSA-N | |
| 82291 | |
| [H+].[Cl-].NCC=C |
| C3H8ClN | |
| MFCD00054329 | |
| 3-Aminopropene Hydrochloride | |
| hydrogen prop-2-en-1-amine chloride |
Safety and Handling
EINECSNumber : (2)-2379
RTECSNumber : BA6950000
TSCA : No