missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allylamine Hydrochloride 98.0+%, TCI America™
$102.52 - $1217.22
Chemical Identifiers
| CAS | 10017-11-5 |
|---|---|
| Molecular Formula | C3H8ClN |
| Molecular Weight (g/mol) | 93.55 |
| MDL Number | MFCD00054329 |
| InChI Key | MLGWTHRHHANFCC-UHFFFAOYSA-N |
| Synonym | 3-Aminopropene Hydrochloride |
| PubChem CID | 82291 |
| IUPAC Name | hydrogen prop-2-en-1-amine chloride |
| SMILES | [H+].[Cl-].NCC=C |
Chemical Identifiers
| 10017-11-5 | |
| 93.55 | |
| MLGWTHRHHANFCC-UHFFFAOYSA-N | |
| 82291 | |
| [H+].[Cl-].NCC=C |
| C3H8ClN | |
| MFCD00054329 | |
| 3-Aminopropene Hydrochloride | |
| hydrogen prop-2-en-1-amine chloride |
Specifications
| 10017-11-5 | |
| C3H8ClN | |
| 25 g | |
| 3-Aminopropene Hydrochloride | |
| [H+].[Cl-].NCC=C | |
| 93.55 | |
| 93.55 | |
| Crystalline Powder |
| 106°C | |
| MFCD00054329 | |
| 2811 | |
| MLGWTHRHHANFCC-UHFFFAOYSA-N | |
| hydrogen prop-2-en-1-amine chloride | |
| 82291 | |
| ≥98.0% (T) | |
| Allylamine Hydrochloride |
Safety and Handling
EINECSNumber : (2)-2379
RTECSNumber : BA6950000
TSCA : No