missing translation for 'onlineSavingsMsg'
Learn More

Allyl vinyl ether, 95% stab. with 0.1% potassium hydroxide

Catalog No. AAL1332009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1332009 10 g
AAL1332018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1332009 Supplier Thermo Scientific Chemicals Supplier No. L1332009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3917-15-5 | C5H8O | 84.12 g/mol

Allyl vinyl ether is used as a intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl vinyl ether is used as a intermediate in chemical research.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, ultraviolet radiation and free radical initiators.

Chemical Identifiers

CAS 3917-15-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00039827
InChI Key ZXABMDQSAABDMG-UHFFFAOYSA-N
Synonym allyl vinyl ether, ether, allyl vinyl, vinyl allyl ether, 1-propene, 3-ethenyloxy, allyl ethenyl ether, unii-fj2j0n55cs, fj2j0n55cs, allylvinylether, acmc-20alul, 3-ethenoxy-1-propene
PubChem CID 221523
IUPAC Name 3-ethenoxyprop-1-ene
SMILES C=CCOC=C

Specifications

Density 0.781
Boiling Point 65°C to 68°C
Flash Point <−5°C (23°F)
Odor Characteristic
Refractive Index 1.412
Linear Formula CH2=CHCH2OCH=CH2
Quantity 10 g
UN Number UN3271
Beilstein 1697665
Solubility Information Slightly soluble in water.
Formula Weight 84.12
Percent Purity 95%
Chemical Name or Material Allyl vinyl ether, stabilized with 0.1% potassium hydroxide
Show More Show Less
Hazard Category H225-H302-H315-H319-H335
Hazard Statement GHS H Statement
H225-H302-H315-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 223-482-6
RTECSNumber KM9450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only