CAS RN 3917-15-5
IUPAC Name:
3-(ethenyloxy)prop-1-ene
Synonyms:
1-propene, 3-(ethenyloxy)-
Molecular Weight (g/mol):
84.12
Molecular Formula:
C5H8O
InChi Key:
ZXABMDQSAABDMG-UHFFFAOYSA-N
SMILES:
C=CCOC=C
1
–
1
of
1
results
1
Promotions Available
Allyl vinyl ether, 95% stab. with 0.1% potassium hydroxide
Allyl vinyl ether, 95% stab. with 0.1% potassium hydroxide
CAS: 3917-15-5 Molecular Formula: C5H8O Molecular Weight (g/mol): 84.12 MDL Number: MFCD00039827 InChI Key: ZXABMDQSAABDMG-UHFFFAOYSA-N Synonym: allyl vinyl ether,ether, allyl vinyl,vinyl allyl ether,1-propene, 3-ethenyloxy,allyl ethenyl ether,unii-fj2j0n55cs,fj2j0n55cs,allylvinylether,acmc-20alul,3-ethenoxy-1-propene PubChem CID: 221523 IUPAC Name: 3-ethenoxyprop-1-ene SMILES: C=CCOC=C