missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl sulfide, ≥97.0% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 592-88-1 |
| Vapor Pressure | 7 mmHg (20°C) |
| Vapor Density | 3.9 (vs. air) |
| Density | 0.887 g/mL (at 25°C) |
| Flash Point | 46°C |
| Physical Form | Neat |
| Linear Formula | (CH2=CHCH2)2 S |
| MDL Number | MFCD00008658 |
| Melting Point | -83°C |
| Boiling Point | 138°C |
| Show More |