missing translation for 'onlineSavingsMsg'
Learn More

Allyl phenyl sulfone, 98%

CAS: 16212-05-8 | C9H10O2S | 182.237 g/mol

$95.16 - $300.10

Chemical Identifiers

CAS 16212-05-8
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00014739
InChI Key KYPIULIVYSQNNT-UHFFFAOYSA-N
Synonym allyl phenyl sulfone, benzene, 2-propenylsulfonyl, allylsulfonyl benzene, phenyl allyl sulfone, sulfone, allyl phenyl, allylsulphonyl benzene, allylphenyl sulfone, 3-phenylsulfonyl-1-propene, benzene, 2-propen-1-ylsulfonyl
PubChem CID 85330
IUPAC Name prop-2-enylsulfonylbenzene
SMILES C=CCS(=O)(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1040106
View Documents
Thermo Scientific Chemicals
A1040106
5 g N/A
Only null left
 
AAA1040114
View Documents
Thermo Scientific Chemicals
A1040114
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16212-05-8
182.237
KYPIULIVYSQNNT-UHFFFAOYSA-N
85330
C=CCS(=O)(=O)C1=CC=CC=C1
C9H10O2S
MFCD00014739
allyl phenyl sulfone, benzene, 2-propenylsulfonyl, allylsulfonyl benzene, phenyl allyl sulfone, sulfone, allyl phenyl, allylsulphonyl benzene, allylphenyl sulfone, 3-phenylsulfonyl-1-propene, benzene, 2-propen-1-ylsulfonyl
prop-2-enylsulfonylbenzene

Specifications

16212-05-8
111°C to 113°C (0.5 mmHg)
C9H10O2S
5 g
Light sensitive
KYPIULIVYSQNNT-UHFFFAOYSA-N
prop-2-enylsulfonylbenzene
85330
98%
1.189
>110°C (230°F)
MFCD00014739
1863561
allyl phenyl sulfone, benzene, 2-propenylsulfonyl, allylsulfonyl benzene, phenyl allyl sulfone, sulfone, allyl phenyl, allylsulphonyl benzene, allylphenyl sulfone, 3-phenylsulfonyl-1-propene, benzene, 2-propen-1-ylsulfonyl
C=CCS(=O)(=O)C1=CC=CC=C1
182.237
182.24
Allyl phenyl sulfone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 240-338-8

RTECSNumber : WR2400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only