missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Phenyl Sulfide 98.0+%, TCI America™
Supplier: TCI America A144225G
Specifications
| Allyl Phenyl Sulfide | |
| Colorless | |
| MFCD00014957 | |
| allyl phenyl sulfide, allylthio benzene, allyl phenyl sulfane, benzene, 2-propenylthio, allylphenyl sulfide, phenyl allyl sulfide, allylphenylthioether, allyl phenyl ;sulfane, sulfide, allyl phenyl | |
| C=CCSC1=CC=CC=C1 | |
| 150.24 | |
| 150.24 | |
| Liquid |
| 5296-64-0 | |
| C9H10S | |
| 25 g | |
| QGNRLAFFKKBSIM-UHFFFAOYSA-N | |
| (prop-2-en-1-ylsulfanyl)benzene | |
| 79180 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 5296-64-0 | |
| 150.24 | |
| QGNRLAFFKKBSIM-UHFFFAOYSA-N | |
| 79180 | |
| C=CCSC1=CC=CC=C1 |
| C9H10S | |
| MFCD00014957 | |
| allyl phenyl sulfide, allylthio benzene, allyl phenyl sulfane, benzene, 2-propenylthio, allylphenyl sulfide, phenyl allyl sulfide, allylphenylthioether, allyl phenyl ;sulfane, sulfide, allyl phenyl | |
| (prop-2-en-1-ylsulfanyl)benzene |
Safety and Handling
TSCA : No