missing translation for 'onlineSavingsMsg'
Learn More

Allyl phenyl sulfide, 97%, Thermo Scientific Chemicals

Catalog No. p-7044134
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0477109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0477109 Supplier Thermo Scientific Chemicals Supplier No. L0477109

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5296-64-0
Molecular Formula C9H10S
Molecular Weight (g/mol) 150.24
MDL Number MFCD00014957
InChI Key QGNRLAFFKKBSIM-UHFFFAOYSA-N
Synonym allyl phenyl sulfide, allylthio benzene, allyl phenyl sulfane, benzene, 2-propenylthio, allylphenyl sulfide, phenyl allyl sulfide, allylphenylthioether, allyl phenyl ;sulfane, sulfide, allyl phenyl
PubChem CID 79180
IUPAC Name prop-2-enylsulfanylbenzene
SMILES C=CCSC1=CC=CC=C1

Specifications

Density 1.024
Boiling Point 223°C to 225°C
Flash Point >98°C (208°F)
Odor Unpleasant
Refractive Index 1.574
Quantity 10 g
Beilstein 1856940
Sensitivity Air Sensitive
Formula Weight 150.24
Percent Purity 97%
Chemical Name or Material Allyl phenyl sulfide
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 226-142-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only