missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Phenoxyacetate 98.0+%, TCI America™
$89.48 - $331.81
Chemical Identifiers
| CAS | 7493-74-5 |
|---|---|
| Molecular Formula | C11H12O3 |
| Molecular Weight (g/mol) | 192.214 |
| MDL Number | MFCD00044632 |
| InChI Key | VUFZVGQUAVDKMC-UHFFFAOYSA-N |
| Synonym | allyl phenoxyacetate, allyl 2-phenoxyacetate, acetic acid, phenoxy-, 2-propenyl ester, acetate p.a., acetate pa, phenoxyacetic acid allyl ester, 2-propenyl phenoxyacetate, unii-q3p8uaf9we, acetic acid, phenoxy-, allyl ester, acetate p.a |
| PubChem CID | 24117 |
| IUPAC Name | prop-2-enyl 2-phenoxyacetate |
| SMILES | C=CCOC(=O)COC1=CC=CC=C1 |
Chemical Identifiers
| 7493-74-5 | |
| 192.214 | |
| VUFZVGQUAVDKMC-UHFFFAOYSA-N | |
| 24117 | |
| C=CCOC(=O)COC1=CC=CC=C1 |
| C11H12O3 | |
| MFCD00044632 | |
| allyl phenoxyacetate, allyl 2-phenoxyacetate, acetic acid, phenoxy-, 2-propenyl ester, acetate p.a., acetate pa, phenoxyacetic acid allyl ester, 2-propenyl phenoxyacetate, unii-q3p8uaf9we, acetic acid, phenoxy-, allyl ester, acetate p.a | |
| prop-2-enyl 2-phenoxyacetate |
Specifications
| 7493-74-5 | |
| 137°C | |
| MFCD00044632 | |
| 2810 | |
| VUFZVGQUAVDKMC-UHFFFAOYSA-N | |
| prop-2-enyl 2-phenoxyacetate | |
| 24117 | |
| ≥98.0% (GC) | |
| Allyl Phenoxyacetate |
| Colorless | |
| C11H12O3 | |
| 25 g | |
| allyl phenoxyacetate, allyl 2-phenoxyacetate, acetic acid, phenoxy-, 2-propenyl ester, acetate p.a., acetate pa, phenoxyacetic acid allyl ester, 2-propenyl phenoxyacetate, unii-q3p8uaf9we, acetic acid, phenoxy-, allyl ester, acetate p.a | |
| C=CCOC(=O)COC1=CC=CC=C1 | |
| 192.214 | |
| 192.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0633
RTECSNumber : AJ2240000
TSCA : Yes