missing translation for 'onlineSavingsMsg'
Learn More

Allyl p-Tolyl Ether 95.0+%, TCI America™

Catalog No. p-7117350
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12875G 5 g
A128725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12875G Supplier TCI America Supplier No. A12875G
Only null left

Chemical Identifiers

CAS 23431-48-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00026098
InChI Key AROCNZZBLCAOPH-UHFFFAOYSA-N
Synonym 4-Allyloxytoluene
PubChem CID 262070
IUPAC Name 1-methyl-4-prop-2-enoxybenzene
SMILES CC1=CC=C(C=C1)OCC=C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 148.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl p-Tolyl Ether